ENA048304205
Enamine
4-imino-5-(4-methyl-1,3-thiazol-2-yl)-2-sulfanyl-3-azaspiro[5.5]undec-1-ene-1-carbonitrile
About This Item
Iupac Name
4-imino-5-(4-methyl-1,3-thiazol-2-yl)-2-sulfanyl-3-azaspiro[5.5]undec-1-ene-1-carbonitrile
Inchi Code
1S/C15H18N4S2/c1-9-8-21-14(18-9)11-12(17)19-13(20)10(7-16)15(11)5-3-2-4-6-15/h8,10H,2-6,17H2,1H3,(H,19,20)
InChI key
KYWCRWGOCMMOCA-UHFFFAOYSA-N
Purity
Country of Origin
Product Link
MSDS
https://www.enaminestore.com/catalog/EN300-11747
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service