ENA048303787
Enamine
3-{[2-(4-sulfamoylphenyl)ethyl]sulfamoyl}benzoic acid
About This Item
mp
198-199
Iupac Name
3-[({2-[4-(aminosulfonyl)phenyl]ethyl}amino)sulfonyl]benzoic acid
Inchi Code
1S/C15H16N2O6S2/c16-24(20,21)13-6-4-11(5-7-13)8-9-17-25(22,23)14-3-1-2-12(10-14)15(18)19/h1-7,10,17H,8-9H2,(H,18,19)(H2,16,20,21)
InChI key
OZSAETMVUPPNPB-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
Melting Point
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-11462
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service