Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA018109962

Enamine

2-[2-(chloromethyl)-4-oxo-3,4-dihydroquinazolin-3-yl]acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
233.66
MDL number:

mp

154-155

Iupac Name

(2-(chloromethyl)-4-oxo-3(4H)-quinazolinyl)acetonitrile

Inchi Code

1S/C11H8ClN3O/c12-7-10-14-9-4-2-1-3-8(9)11(16)15(10)6-5-13/h1-4,7,14H,6H2

InChI key

RRPBAEQMGYBZHN-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-06885

Melting Point

154-155

MSDS

https://www.enaminestore.com/catalog/EN300-06885


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service