Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA018107775

Enamine

3-methoxy-4-(2-methylpropoxy)benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$24.70
100 MG
$24.70
250 MG
$24.70
500 MG
$24.70
2.5 G
$36.40
5 G
$65.00
10 G
$117.00

Vendor SKU: EN300-04350-1G

$24.70


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
1 G
$24.70
100 MG
$24.70
250 MG
$24.70
500 MG
$24.70
2.5 G
$36.40
5 G
$65.00
10 G
$117.00

About This Item

CAS Number:
Molecular Weight:
224.26
MDL number:

Vendor SKU: EN300-04350-1G

$24.70


Ships in 7 days - from Aldrich Partner.

mp

142-143

Iupac Name

4-isobutoxy-3-methoxybenzoic acid

Inchi Code

1S/C12H16O4/c1-8(2)7-16-10-5-4-9(12(13)14)6-11(10)15-3/h4-6,8H,7H2,1-3H3,(H,13,14)

InChI key

SBHFJHQPZDLFTD-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-04350

Melting Point

142-143

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-04350


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service