Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CO1H324A9947

CombiPhos Catalysts Inc

tert-butyl 3-(6-bromopyridin-3-yl)pyrrolidine-1-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$750.00
100 MG
$1,247.50
250 MG
$2,250.00
1 G
$4,375.00

Vendor SKU: CS925-50MG

$750.00


Ships in 30 days - from Aldrich Partner.


Select a Size

Change View
50 MG
$750.00
100 MG
$1,247.50
250 MG
$2,250.00
1 G
$4,375.00

About This Item

CAS Number:
Molecular Weight:
327.22

Vendor SKU: CS925-50MG

$750.00


Ships in 30 days - from Aldrich Partner.

Iupac Name

tert-butyl 3-(6-bromopyridin-3-yl)pyrrolidine-1-carboxylate

Inchi Code

1S/C14H19BrN2O2/c1-14(2,3)19-13(18)17-7-6-11(9-17)10-4-5-12(15)16-8-10/h4-5,8,11H,6-7,9H2,1-3H3

InChI key

IEDPLPSTQPREPW-UHFFFAOYSA-N

Purity

95%

Country of Origin

US

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service