Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CO1H324A9786

CombiPhos Catalysts Inc

3-(1,4-dioxan-2-yl)aniline

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$475.00
100 MG
$1,247.50
250 MG
$1,875.00
1 G
$3,125.00
5 G
$6,875.00

Vendor SKU: CS7961-N-25MG

$475.00


Ships in 30 days - from Aldrich Partner.


Select a Size

Change View
25 MG
$475.00
100 MG
$1,247.50
250 MG
$1,875.00
1 G
$3,125.00
5 G
$6,875.00

About This Item

CAS Number:
Molecular Weight:
179.22

Vendor SKU: CS7961-N-25MG

$475.00


Ships in 30 days - from Aldrich Partner.

Iupac Name

3-(1,4-dioxan-2-yl)aniline

Inchi Code

1S/C10H13NO2/c11-9-3-1-2-8(6-9)10-7-12-4-5-13-10/h1-3,6,10H,4-5,7,11H2

InChI key

FMLFIUBUHUSOAT-UHFFFAOYSA-N

Purity

95%

Country of Origin

US

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service