CO1H324A801A
CombiPhos Catalysts Inc
4-Amino-2-(2-methoxyphenyl)pyrimidine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Molecular Weight:
201.23
Iupac Name
2-(2-methoxyphenyl)-4-pyrimidinamine
Inchi Code
1S/C11H11N3O/c1-15-9-5-3-2-4-8(9)11-13-7-6-10(12)14-11/h2-7H,1H3,(H2,12,13,14)
InChI key
XCKMKYFZZNDBHC-UHFFFAOYSA-N
Purity
95%
Country of Origin
US
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service