Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COMH580D81CF

Combi-Blocks, Inc.

2-[2-(benzyloxy)ethoxy]-1,4-dibromobenzene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
386.08
MDL number:

Iupac Name

2-(2-(benzyloxy)ethoxy)-1,4-dibromobenzene

Inchi Code

1S/C15H14Br2O2/c16-13-6-7-14(17)15(10-13)19-9-8-18-11-12-4-2-1-3-5-12/h1-7,10H,8-9,11H2

InChI key

VXVVTKFPLJWTJN-UHFFFAOYSA-N

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?OT-5042

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?OT-5042


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service