Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COMH4211B1C4

Combi-Blocks, Inc.

4-(2,2-Difluoroethoxy)-2-fluorobenzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$910.00
5 G
$2,730.00

Vendor SKU: QD-4065-1G

$910.00


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
1 G
$910.00
5 G
$2,730.00

About This Item

CAS Number:
Molecular Weight:
204.15
MDL number:

Vendor SKU: QD-4065-1G

$910.00


Ships in 7 days - from Aldrich Partner.

Iupac Name

4-(2,2-difluoroethoxy)-2-fluorobenzaldehyde

Inchi Code

1S/C9H7F3O2/c10-8-3-7(14-5-9(11)12)2-1-6(8)4-13/h1-4,9H,5H2

InChI key

QCZKVWQAILMHSH-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QD-4065

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QD-4065


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service