Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COMH042390F1

Combi-Blocks, Inc.

2-Ethoxy-4-formylphenyl thiophene-2-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$260.00
5 G
$910.00

Vendor SKU: QZ-8357-1G

$260.00


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
1 G
$260.00
5 G
$910.00

About This Item

CAS Number:
Molecular Weight:
276.31
MDL number:

Vendor SKU: QZ-8357-1G

$260.00


Ships in 7 days - from Aldrich Partner.

Iupac Name

2-ethoxy-4-formylphenyl 2-thiophenecarboxylate

Inchi Code

1S/C14H12O4S/c1-2-17-12-8-10(9-15)5-6-11(12)18-14(16)13-4-3-7-19-13/h3-9H,2H2,1H3

InChI key

XWUSMUFRTPJYOJ-UHFFFAOYSA-N

storage temp.

Refrigerated

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8357

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8357


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service