COMH04237CDF
Combi-Blocks, Inc.
4-Hydroxybenzaldehyde (5-isopropyl-4-oxo-4,5-dihydro-1,3-thiazol-2-yl)hydrazone
About This Item
Iupac Name
4-hydroxybenzaldehyde [(2E)-5-isopropyl-4-oxo-1,3-thiazolidin-2-ylidene]hydrazone
Inchi Code
1S/C13H15N3O2S/c1-8(2)11-12(18)15-13(19-11)16-14-7-9-3-5-10(17)6-4-9/h3-8,17-18H,1-2H3,(H,15,16)/b14-7+
Inchi Key
REWWVGLNJAXWTM-VGOFMYFVSA-N
Storage temp.
Purity
Country of Origin
Product Link
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3143
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service