Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COM497526088

Combi-Blocks, Inc.

1-(Azetidin-3-yl)azetidin-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
128.17
MDL number:

Iupac Name

[1,3'-biazetidin]-3-ol

Inchi Code

1S/C6H12N2O/c9-6-3-8(4-6)5-1-7-2-5/h5-7,9H,1-4H2

InChI key

JAPZEBGUFUANNT-UHFFFAOYSA-N

storage temp.

Refrigerated

Purity

97%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QM-7972

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QM-7972


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service