Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COM448645981

Combi-Blocks, Inc.

D-Fructose-1,6-diphosphate dicalcium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
416.24
MDL number:

Iupac Name

dicalcium 1,6-di-O-phosphonato-D-fructose

Inchi Code

1S/C6H14O12P2.2Ca/c7-3(1-17-19(11,12)13)5(9)6(10)4(8)2-18-20(14,15)16;;/h3,5-7,9-10H,1-2H2,(H2,11,12,13)(H2,14,15,16);;/q;2*+2/p-4/t3-,5-,6-;;/m1../s1

InChI key

MKSZTTCFYSGQHA-ABICQQBESA-J

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QB-1767

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QB-1767


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service