Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COM448622904

Combi-Blocks, Inc.

Benzyl N-[(1S)-1-(propylcarbamoyl)ethyl]carbamate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$78.00
5 G
$195.00
25 G
$416.00
100 G
$1,092.00

Vendor SKU: LD-0834-1G

$78.00


Ships in 7 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 G
$78.00
5 G
$195.00
25 G
$416.00
100 G
$1,092.00

About This Item

CAS Number:
Molecular Weight:
264.32
MDL number:

Vendor SKU: LD-0834-1G

$78.00


Ships in 7 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

benzyl (1S)-1-methyl-2-oxo-2-(propylamino)ethylcarbamate

Inchi Code

1S/C14H20N2O3/c1-3-9-15-13(17)11(2)16-14(18)19-10-12-7-5-4-6-8-12/h4-8,11H,3,9-10H2,1-2H3,(H,15,17)(H,16,18)/t11-/m0/s1

InChI key

QTULOXLTYMFRQZ-NSHDSACASA-N

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?LD-0834

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?LD-0834


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service