COM448622406
Combi-Blocks, Inc.
4-[3-(Trifluoromethyl)phenyl]benzonitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
3'-(trifluoromethyl)[1,1'-biphenyl]-4-carbonitrile
Inchi Code
1S/C14H8F3N/c15-14(16,17)13-3-1-2-12(8-13)11-6-4-10(9-18)5-7-11/h1-8H
InChI key
OCPLIDYXKIIHKX-UHFFFAOYSA-N
Purity
98%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?LD-0032
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?LD-0032
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service