Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COM448620032

Combi-Blocks, Inc.

1-Bromo-3-(ethanesulfinyl)benzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$104.00
5 G
$312.00

Vendor SKU: HC-3596-1G

$104.00


Availability
Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
1 G
$104.00
5 G
$312.00

About This Item

CAS Number:
Molecular Weight:
233.13
MDL number:

Vendor SKU: HC-3596-1G

$104.00


Availability
Ships in 7 days - from Aldrich Partner.

Iupac Name

1-bromo-3-(ethylsulfinyl)benzene

Inchi Code

1S/C8H9BrOS/c1-2-11(10)8-5-3-4-7(9)6-8/h3-6H,2H2,1H3

InChI key

KTAYYVGVOVUOOL-UHFFFAOYSA-N

Purity

98%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?HC-3596

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?HC-3596


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service