Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COM448620028

Combi-Blocks, Inc.

Methyl 2-methyl-2-[4-(trifluoromethyl)phenyl]propanoate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$208.00
5 G
$624.00
10 G
$1,092.00

Vendor SKU: HC-3591-1G

$208.00


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
1 G
$208.00
5 G
$624.00
10 G
$1,092.00

About This Item

CAS Number:
Molecular Weight:
246.23
MDL number:

Vendor SKU: HC-3591-1G

$208.00


Ships in 7 days - from Aldrich Partner.

Iupac Name

methyl 2-methyl-2-(4-(trifluoromethyl)phenyl)propanoate

Inchi Code

1S/C12H13F3O2/c1-11(2,10(16)17-3)8-4-6-9(7-5-8)12(13,14)15/h4-7H,1-3H3

InChI key

ZIIRSEIPZKDSQH-UHFFFAOYSA-N

Purity

98%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?HC-3591

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?HC-3591


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service