Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COM448616391

Combi-Blocks, Inc.

4-Bis(N-Boc)amino-3-fluorophenylboronic acid

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
355.17
MDL number:

Iupac Name

(4-(bis(tert-butoxycarbonyl)amino)-3-fluorophenyl)boronic acid

Inchi Code

1S/C16H23BFNO6/c1-15(2,3)24-13(20)19(14(21)25-16(4,5)6)12-8-7-10(17(22)23)9-11(12)18/h7-9,22-23H,1-6H3

InChI key

PDOXRNGFOOHMLN-UHFFFAOYSA-N

storage temp.

Refrigerated

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?BC-1021

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?BC-1021


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service