Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COM448614803

Combi-Blocks, Inc.

3-Bromo-2-fluoro-5-(trifluoromethoxy)phenylboronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$195.00
1 G
$455.00
5 G
$1,274.00

Vendor SKU: BB-4491-250MG

$195.00


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$195.00
1 G
$455.00
5 G
$1,274.00

About This Item

CAS Number:
Molecular Weight:
302.82
MDL number:

Vendor SKU: BB-4491-250MG

$195.00


Ships in 7 days - from Aldrich Partner.

Iupac Name

3-bromo-2-fluoro-5-(trifluoromethoxy)phenylboronic acid

Inchi Code

1S/C7H4BBrF4O3/c9-5-2-3(16-7(11,12)13)1-4(6(5)10)8(14)15/h1-2,14-15H

InChI key

IPONDPCBQLMVCI-UHFFFAOYSA-N

storage temp.

Refrigerated

Purity

98%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?BB-4491

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?BB-4491


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service