COM448613382
Combi-Blocks, Inc.
4-[4-(Dimethylsulfamoyl)piperazin-1-yl]phenylboronic acid
About This Item
Iupac Name
(4-(4-(N,N-dimethylsulfamoyl)piperazin-1-yl)phenyl)boronic acid
Inchi Code
1S/C12H20BN3O4S/c1-14(2)21(19,20)16-9-7-15(8-10-16)12-5-3-11(4-6-12)13(17)18/h3-6,17-18H,7-10H2,1-2H3
InChI key
PJSBLYFEFXMCNH-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Product Link
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?BB-0695
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service