COM448613124
Combi-Blocks, Inc.
(7-Chloro-8-methylquinolin-6-yl)boronic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
(7-chloro-8-methylquinolin-6-yl)boronic acid
Inchi Code
1S/C10H9BClNO2/c1-6-9(12)8(11(14)15)5-7-3-2-4-13-10(6)7/h2-5,14-15H,1H3
InChI key
RDJIHCAVLJQCFP-UHFFFAOYSA-N
storage temp.
Refrigerated
Purity
95%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?BB-0402
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?BB-0402
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service