Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CIAH98D1EAA6

ChemScene LLC (Preferred Partner)

Dodecylbenzenesulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$60.00

Vendor SKU: CS-0256295-1KG

$60.00


Ships in 7 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 KG
$60.00

About This Item

CAS Number:
Molecular Weight:
326.5
MDL number:

Vendor SKU: CS-0256295-1KG

$60.00


Ships in 7 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

bp

82 C at 760 mmHg

mp

10 C

Iupac Name

2-dodecylbenzenesulfonic acid

Inchi Code

1S/C18H30O3S/c1-2-3-4-5-6-7-8-9-10-11-14-17-15-12-13-16-18(17)22(19,20)21/h12-13,15-16H,2-11,14H2,1H3,(H,19,20,21)

InChI key

WBIQQQGBSDOWNP-UHFFFAOYSA-N

storage temp.

4C, sealed storage, away from moisture

Purity

80%

Country of Origin

CN

Physical Form

Liquid

Product Link

https://www.chemscene.com/27176-87-0.html

Boiling point

82 C at 760 mmHg

Melting Point

10 C

Signal Word

Danger

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0256295.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service