Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CIAH987F5121

ChemScene LLC (Preferred Partner)

2-Fluoro-5-formylbenzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$20.00

Vendor SKU: CS-M3441-1G

$20.00


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
1 G
$20.00

About This Item

CAS Number:
Molecular Weight:
168.12
MDL number:

Vendor SKU: CS-M3441-1G

$20.00


Ships in 7 days - from Aldrich Partner.

bp

330.9C at 760 mmHg

mp

175-178C

Iupac Name

2-fluoro-5-formylbenzoic acid

Inchi Code

1S/C8H5FO3/c9-7-2-1-5(4-10)3-6(7)8(11)12/h1-4H,(H,11,12)

InChI key

XZUFXXPSLGVLFC-UHFFFAOYSA-N

storage temp.

4C

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/550363-85-4.html

Flash Point

153.9C

Boiling point

330.9C at 760 mmHg

Melting Point

175-178C

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-M3441.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service