Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CIAH987EFC39

ChemScene LLC (Preferred Partner)

3-Bromobenzhydrazide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$30.00

Vendor SKU: CS-W014905-25G

$30.00


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
25 G
$30.00

About This Item

CAS Number:
Molecular Weight:
215.05
MDL number:

Vendor SKU: CS-W014905-25G

$30.00


Ships in 7 days - from Aldrich Partner.

bp

368.5C at 760 mmHg

mp

155-156C

Iupac Name

3-bromobenzohydrazide

Inchi Code

1S/C7H7BrN2O/c8-6-3-1-2-5(4-6)7(11)10-9/h1-4H,9H2,(H,10,11)

InChI key

BNAQRAZIPAHWAR-UHFFFAOYSA-N

storage temp.

Store at room temperature

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/39115-96-3.html

Boiling point

368.5C at 760 mmHg

Melting Point

155-156C

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-W014905.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service