Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CIAH9917D1AC

ChemScene LLC (Preferred Partner)

tert-Butyl (S)-(3-oxobutan-2-yl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
187.24
MDL number:

bp

270.7+-23.0 C at 760 mmHg

mp

36-38 C

Iupac Name

tert-butyl (1S)-1-methyl-2-oxopropylcarbamate

Inchi Code

1S/C9H17NO3/c1-6(7(2)11)10-8(12)13-9(3,4)5/h6H,1-5H3,(H,10,12)/t6-/m0/s1

Inchi Key

NUAKKRMYBBXGFP-LURJTMIESA-N

Storage temp.

Store at room temperature

Purity

96%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/126027-07-4.html

Boiling point

270.7+-23.0 C at 760 mmHg

Melting Point

36-38 C

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0608799.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service