Skip to Content
MilliporeSigma
All Photos(1)

Documents

CIAH98D1F873

ChemScene LLC (Preferred Partner)

tert-Butyl 4-(2-aminophenyl)piperidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
276.38
MDL number:

Iupac Name

tert-butyl 4-(2-aminophenyl)-1-piperidinecarboxylate

Inchi Code

1S/C16H24N2O2/c1-16(2,3)20-15(19)18-10-8-12(9-11-18)13-6-4-5-7-14(13)17/h4-7,12H,8-11,17H2,1-3H3

InChI key

BSLRAYZVIBFRQQ-UHFFFAOYSA-N

storage temp.

4C, protect from light

Purity

98%

Country of Origin

CN

Product Link

https://www.chemscene.com/199105-03-8.html

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service