Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CIAH98D1F73D

ChemScene LLC (Preferred Partner)

[2-chloro-5-(trifluoromethyl)phenyl]hydrazine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 G
$60.00
5 G
$118.00

Vendor SKU: CS-0219263-2.5G

$60.00


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
2.5 G
$60.00
5 G
$118.00

About This Item

CAS Number:
Molecular Weight:
247.05
MDL number:

Vendor SKU: CS-0219263-2.5G

$60.00


Ships in 7 days - from Aldrich Partner.

Iupac Name

1-[2-chloro-5-(trifluoromethyl)phenyl]hydrazine hydrochloride

Inchi Code

1S/C7H6ClF3N2.ClH/c8-5-2-1-4(7(9,10)11)3-6(5)13-12;/h1-3,13H,12H2;1H

InChI key

FRLINAXMXPSHSI-UHFFFAOYSA-N

storage temp.

4C, stored under nitrogen

Salt Data

Hydrochloride

Purity

97%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/299163-40-9.html

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0219263.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service