Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CIAH98D1EA62

ChemScene LLC (Preferred Partner)

2,2-Diphenyloxirane

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$76.00
1 G
$212.00
5 G
$496.00

Vendor SKU: CS-0319255-250MG

$76.00


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$76.00
1 G
$212.00
5 G
$496.00

About This Item

CAS Number:
Molecular Weight:
196.25
MDL number:

Vendor SKU: CS-0319255-250MG

$76.00


Ships in 7 days - from Aldrich Partner.

bp

140-142 C at 5 mmHg

mp

54-56 C

Iupac Name

2,2-diphenyloxirane

Inchi Code

1S/C14H12O/c1-3-7-12(8-4-1)14(11-15-14)13-9-5-2-6-10-13/h1-10H,11H2

InChI key

PRLJMHVNHLTQJJ-UHFFFAOYSA-N

storage temp.

4C

Purity

95%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/882-59-7.html

Boiling point

140-142 C at 5 mmHg

Melting Point

54-56 C

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0319255.pdf


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service