Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CIAH98D1E985

ChemScene LLC (Preferred Partner)

1-(3-(Triethoxysilyl)propyl)-1H-pyrrole-2,5-dione

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$17.00
5 G
$71.00
25 G
$251.00

Vendor SKU: CS-0161834-1G

$17.00


Availability
Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
1 G
$17.00
5 G
$71.00
25 G
$251.00

About This Item

CAS Number:
Molecular Weight:
301.41
MDL number:

Vendor SKU: CS-0161834-1G

$17.00


Availability
Ships in 7 days - from Aldrich Partner.

bp

350.2+-25.0 C at 760 mmHg

mp

N/A

density

1.099+-0.06 g/cm3

Iupac Name

1-(3-(triethoxysilyl)propyl)-1H-pyrrole-2,5-dione

Inchi Code

1S/C13H23NO5Si/c1-4-17-20(18-5-2,19-6-3)11-7-10-14-12(15)8-9-13(14)16/h8-9H,4-7,10-11H2,1-3H3

InChI key

YPKNIURCMPHHJJ-UHFFFAOYSA-N

storage temp.

4C

Purity

95%

Country of Origin

CN

Physical Form

Solid-Liquid Mixture

Product Link

https://www.chemscene.com/29602-11-7.html

Boiling point

350.2+-25.0 C at 760 mmHg

Melting Point

N/A

density

1.099+-0.06 g/cm3

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0161834.pdf


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service