Skip to Content
MilliporeSigma
All Photos(1)

Documents

CIAH98B92C49

ChemScene LLC (Preferred Partner)

Di-tert-butyl(neopentyl)phosphonium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
304.16
MDL number:

Iupac Name

di-tert-butyl(2,2-dimethylpropyl)phosphane trifluoroborane hydrofluoride

Inchi Code

1S/C13H29P.BF3.FH/c1-11(2,3)10-14(12(4,5)6)13(7,8)9;2-1(3)4;/h10H2,1-9H3;;1H

InChI key

IKIGXHALFGJPNB-UHFFFAOYSA-N

storage temp.

4C, protect from light, stored under nitrogen

Salt Data

Tetrafluoroborate

Purity

97%

Country of Origin

CN

Product Link

https://www.chemscene.com/886059-84-3.html

Signal Word

Warning

Precautionary statements


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service