Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CIAH98ABF93F

ChemScene LLC (Preferred Partner)

4-Methylbenzo[b]thiophene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
148.23
MDL number:

bp

243C at 760 mmHg

mp

-6--4 C

Iupac Name

4-methyl-1-benzothiophene

Inchi Code

1S/C9H8S/c1-7-3-2-4-9-8(7)5-6-10-9/h2-6H,1H3

InChI key

RPKWIZPGQZKQKY-UHFFFAOYSA-N

storage temp.

4C, protect from light

Purity

98%

Country of Origin

CN

Physical Form

Liquid

Product Link

https://www.chemscene.com/14315-11-8.html

Boiling point

243C at 760 mmHg

Melting Point

-6--4 C

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-W022216.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service