Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CIAH988500DE

ChemScene LLC (Preferred Partner)

(3-Bromo-4-chlorophenyl)hydrazine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
221.48
MDL number:

Iupac Name

(3-bromo-4-chlorophenyl)hydrazine

Inchi Code

1S/C6H6BrClN2/c7-5-3-4(10-9)1-2-6(5)8/h1-3,10H,9H2

InChI key

IZGXAWPNPDVOFB-UHFFFAOYSA-N

storage temp.

4C, stored under nitrogen

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/344248-15-3.html

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0435683.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service