Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CIAH987F2388

ChemScene LLC (Preferred Partner)

tert-Butyl (2-oxoazepan-3-yl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
228.29
MDL number:

bp

417.7+-34.0 C at 760 mmHg

Iupac Name

tert-butyl 2-oxo-3-azepanylcarbamate

Inchi Code

1S/C11H20N2O3/c1-11(2,3)16-10(15)13-8-6-4-5-7-12-9(8)14/h8H,4-7H2,1-3H3,(H,12,14)(H,13,15)

InChI key

AQKNKAUJTJFUMG-UHFFFAOYSA-N

storage temp.

-20C

Purity

97%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/179686-45-4.html

Boiling point

417.7+-34.0 C at 760 mmHg

Signal Word

Warning

Precautionary statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0112893.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service