Skip to Content
MilliporeSigma
All Photos(1)

Documents

CIAH987EC779

ChemScene LLC (Preferred Partner)

2-Bromothiophene-3-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
188.05
MDL number:

bp

284.3+-25.0 C at 760 mmHg

mp

No data available

Iupac Name

2-bromo-3-thiophenecarbonitrile

Inchi Code

1S/C5H2BrNS/c6-5-4(3-7)1-2-8-5/h1-2H

InChI key

GHMDOVNPFWGFSZ-UHFFFAOYSA-N

storage temp.

4C, stored under nitrogen

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/56182-43-5.html

Boiling point

284.3+-25.0 C at 760 mmHg

Melting Point

No data available

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0096797.pdf


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service