Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CIAH987EC2B7

ChemScene LLC (Preferred Partner)

(1S,2S)-N,N-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
662.79
MDL number:

bp

749.8+-60.0 C at 760 mmHg

mp

54-56 C

Iupac Name

(1S,2S)-N1,N2-bis(2-(diphenylphosphanyl)benzyl)cyclohexane-1,2-diamine

Inchi Code

1S/C44H44N2P2/c1-5-21-37(22-6-1)47(38-23-7-2-8-24-38)43-31-17-13-19-35(43)33-45-41-29-15-16-30-42(41)46-34-36-20-14-18-32-44(36)48(39-25-9-3-10-26-39)40-27-11-4-12-28-40/h1-14,17-28,31-32,41-42,45-46H,15-16,29-30,33-34H2/t41-,42-/m0/s1

InChI key

OBHPYVNBXWUKNY-COCZKOEFSA-N

storage temp.

4C, protect from light, stored under nitrogen

Purity

95%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/174677-83-9.html

Boiling point

749.8+-60.0 C at 760 mmHg

Melting Point

54-56 C

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0085215.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service