Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CIAH987EBAEA

ChemScene LLC (Preferred Partner)

Methyl 3-methylpiperidine-4-carboxylate hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$80.00

Vendor SKU: CS-0049072-100MG

$80.00


Availability
Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
100 MG
$80.00

About This Item

CAS Number:
Molecular Weight:
193.67
MDL number:

Vendor SKU: CS-0049072-100MG

$80.00


Availability
Ships in 7 days - from Aldrich Partner.

Iupac Name

methyl 3-methylpiperidine-4-carboxylate hydrochloride

Inchi Code

1S/C8H15NO2.ClH/c1-6-5-9-4-3-7(6)8(10)11-2;/h6-7,9H,3-5H2,1-2H3;1H

InChI key

LIBNHIYBEVBSKI-UHFFFAOYSA-N

storage temp.

Store at room temperature

Salt Data

Hydrochloride

Purity

97%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/1797255-52-7.html

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0049072.pdf


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service