Skip to Content
MilliporeSigma

CH6H9A56DEA2

Chem Impex International, Inc.

(R)-4-(Fmoc-amino)-3-(Z-amino)butyric acid

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
474.51
MDL number:

mp

169-178 C

Iupac Name

4-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(((benzyloxy)carbonyl)amino)butanoic acid

Inchi Code

1S/C27H26N2O6/c30-25(31)14-19(29-27(33)34-16-18-8-2-1-3-9-18)15-28-26(32)35-17-24-22-12-6-4-10-20(22)21-11-5-7-13-23(21)24/h1-13,19,24H,14-17H2,(H,28,32)(H,29,33)(H,30,31)

InChI key

GNBVOVLCIBZKIQ-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

98-101%

Shipping Temperature

RT

Physical Form

White crystalline powder

Product Link

https://www.chemimpex.com/product/productinfo/-i-r-i-4-fmoc-amino-3-z-amino-butyric-acid/20206?cid=1

Melting Point

169-178 C

MSDS

https://www.chemimpex.com/product/productinfo/-i-r-i-4-fmoc-amino-3-z-amino-butyric-acid/20206?cid=1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service