Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CH6H9A56DA3B

Chem Impex International, Inc.

3-(2-Methylphenyl)-1H-pyrazol-5-amine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
173.22
MDL number:

bp

108-112 C / 5 mmHg

Iupac Name

5-(2-methylphenyl)-1H-pyrazol-3-amine

Inchi Code

1S/C10H11N3/c1-7-4-2-3-5-8(7)9-6-10(11)13-12-9/h2-6H,1H3,(H3,11,12,13)

InChI key

CKQCUZKXAXOTEZ-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

98%

Country of Origin

US

Shipping Temperature

RT

Physical Form

Light brown liquid

Product Link

https://www.chemimpex.com/product/productinfo/3-2-methylphenyl-1-i-h-i-pyrazol-5-amine/15294?cid=0

Boiling point

108-112 C / 5 mmHg

MSDS

https://www.chemimpex.com/product/productinfo/3-2-methylphenyl-1-i-h-i-pyrazol-5-amine/15294?cid=0


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service