Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CH6447917446

Chem Impex International, Inc.

2-[4-(4-Fluorophenyl)Phenyl]Ethylamine Hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
251.73
MDL number:

Iupac Name

2-(4'-fluoro[1,1'-biphenyl]-4-yl)ethanamine hydrochloride

Inchi Code

1S/C14H14FN.ClH/c15-14-7-5-13(6-8-14)12-3-1-11(2-4-12)9-10-16;/h1-8H,9-10,16H2;1H

InChI key

FQLJVLOQYYHIIV-UHFFFAOYSA-N

storage temp.

Store at 0-8C

Purity

98%

Shipping Temperature

RT

Physical Form

Pink solid powder

Product Link

https://www.chemimpex.com/product/productinfo/2-4-4-fluorophenyl-phenyl-ethylamine-hydrochloride/13974?cid=0

MSDS

https://www.chemimpex.com/product/productinfo/2-4-4-fluorophenyl-phenyl-ethylamine-hydrochloride/13974?cid=0


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service