Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CH6371387542

Chem Impex International, Inc.

7-Chloro-3,4-dihydro-2H-benzo[b]oxepin-5-one

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
196.63
MDL number:

bp

98-122 C / 5mm

Iupac Name

7-chloro-3,4-dihydro-1-benzoxepin-5(2H)-one

Inchi Code

1S/C10H9ClO2/c11-7-3-4-10-8(6-7)9(12)2-1-5-13-10/h3-4,6H,1-2,5H2

InChI key

SWNUBBODNFTNPC-UHFFFAOYSA-N

storage temp.

Store at 0-8C

Purity

97%

Country of Origin

US

Shipping Temperature

RT

Physical Form

Colorless to yellow liquid

Product Link

https://www.chemimpex.com/product/productinfo/7-chloro-3-4-dihydro-2-i-h-i-benzo-i-beta-i-oxepin-5-one/16410?cid=0

Boiling point

98-122 C / 5mm

MSDS

https://www.chemimpex.com/product/productinfo/7-chloro-3-4-dihydro-2-i-h-i-benzo-i-beta-i-oxepin-5-one/16410?cid=0


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service