CH6371380650
Chem Impex International, Inc.
Z-DL-leucine amide
About This Item
mp
129-135 C
Iupac Name
benzyl 1-(aminocarbonyl)-3-methylbutylcarbamate
Inchi Code
1S/C14H20N2O3/c1-10(2)8-12(13(15)17)16-14(18)19-9-11-6-4-3-5-7-11/h3-7,10,12H,8-9H2,1-2H3,(H2,15,17)(H,16,18)
InChI key
JZEYMGMOBIYUOV-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Shipping Temperature
Product Link
Melting Point
MSDS
https://www.chemimpex.com/product/productinfo/z-dl-leucine-amide/5736?cid=0
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service