Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C12H98822448

Chemcia Scientific, LLC

4-[(2,2-Dimethyl-4,6-dioxo-[1,3]dioxan-5-ylidenemethyl)-amino]-benzoic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
319.31
MDL number:

Iupac Name

ethyl 4-{[(2,2-dimethyl-4,6-dioxo-1,3-dioxan-5-ylidene)methyl]amino}benzoate

Inchi Code

1S/C16H17NO6/c1-4-21-13(18)10-5-7-11(8-6-10)17-9-12-14(19)22-16(2,3)23-15(12)20/h5-9,17H,4H2,1-3H3

InChI key

ZMPTVSZNLHVTRO-UHFFFAOYSA-N

storage temp.

0-5C

Purity

97%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

Yellow Crystalline Solid

Product Link

http://www.chemcia.com/product_fl02.asp?id=450

MSDS

http://www.chemcia.com/product_fl02.asp?id=450


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service