Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C12H961E9DBC

Chemcia Scientific, LLC

2-Phenyl-2,4,6,7-tetrahydro-pyrazolo[4,3-c]pyridine-5-carboxylic acid tert-butyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$114.00
1 G
$282.00
5 G
$870.00

Vendor SKU: CC00-6817-250MG

$114.00


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$114.00
1 G
$282.00
5 G
$870.00

About This Item

CAS Number:
Molecular Weight:
299.37
MDL number:

Vendor SKU: CC00-6817-250MG

$114.00


Ships in 7 days - from Aldrich Partner.

Iupac Name

tert-butyl 2-phenyl-2,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate

Inchi Code

1S/C17H21N3O2/c1-17(2,3)22-16(21)19-10-9-15-13(11-19)12-20(18-15)14-7-5-4-6-8-14/h4-8,12H,9-11H2,1-3H3

InChI key

ICNBNCIXLIGCGZ-UHFFFAOYSA-N

storage temp.

0-5C

Purity

97%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

Orange Liquid

Product Link

http://www.chemcia.com/product_fl02.asp?id=3759

MSDS

http://www.chemcia.com/product_fl02.asp?id=3759


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service