Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C12H316CF638

Chemcia Scientific, LLC

5-Fluoro-2H-indazole-3-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
194.17

Iupac Name

methyl 5-fluoro-2H-indazole-3-carboxylate

Inchi Code

1S/C9H7FN2O2/c1-14-9(13)8-6-4-5(10)2-3-7(6)11-12-8/h2-4H,1H3,(H,11,12)

InChI key

YBMSTQBPEVBPRZ-UHFFFAOYSA-N

storage temp.

0-5C

Purity

97%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

Brown Solid

Product Link

http://www.chemcia.com/product_fl02.asp?id=13704

MSDS

http://www.chemcia.com/product_fl02.asp?id=13704


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service