C12H316CF547
Chemcia Scientific, LLC
4-Bromo-2-iodo-5-trifluoromethyl-phenol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Molecular Weight:
366.9
Iupac Name
4-bromo-2-iodo-5-(trifluoromethyl)phenol
Inchi Code
1S/C7H3BrF3IO/c8-4-2-5(12)6(13)1-3(4)7(9,10)11/h1-2,13H
InChI key
VBJQBZQTYNCOGD-UHFFFAOYSA-N
storage temp.
0-5C
Purity
95%
Country of Origin
US
Shipping Temperature
Ambient
Physical Form
Yellow Liquid
Product Link
http://www.chemcia.com/product_fl02.asp?id=10871
MSDS
http://www.chemcia.com/product_fl02.asp?id=10871
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service