Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C12H316CED2D

Chemcia Scientific, LLC

2,6-Dibromo-[1,2,4]triazolo[1,5-a]pyrazine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
277.91
MDL number:

Iupac Name

2,6-dibromo-[1,2,4]triazolo[1,5-a]pyrazine

Inchi Code

1S/C5H2Br2N4/c6-3-2-11-4(1-8-3)9-5(7)10-11/h1-2H

InChI key

BAJUHIQABQGBLO-UHFFFAOYSA-N

storage temp.

0-5

Purity

95%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

White Solid

Product Link

http://www.chemcia.com/product_fl02.asp?id=4295

MSDS

http://www.chemcia.com/product_fl02.asp?id=4295


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service