Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C12H316CEA9E

Chemcia Scientific, LLC

2-Chloro-4,5,6,7-tetrahydro-benzothiazole-6-carboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
245.73
MDL number:

Iupac Name

ethyl 2-chloro-4,5,6,7-tetrahydrobenzo[d]thiazole-6-carboxylate

Inchi Code

1S/C10H12ClNO2S/c1-2-14-9(13)6-3-4-7-8(5-6)15-10(11)12-7/h6H,2-5H2,1H3

InChI key

DRHJCGYWKNJZPL-UHFFFAOYSA-N

storage temp.

0-5C

Purity

95%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

Brown Liquid

Product Link

http://www.chemcia.com/product_fl02.asp?id=2961

MSDS

http://www.chemcia.com/product_fl02.asp?id=2961


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service