C12H316CE9B3
Chemcia Scientific, LLC
4-Cyclobutoxy-benzoic acid methyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 4-cyclobutoxybenzoate
Inchi Code
1S/C12H14O3/c1-14-12(13)9-5-7-11(8-6-9)15-10-3-2-4-10/h5-8,10H,2-4H2,1H3
InChI key
JUWRYQAKTOPCBU-UHFFFAOYSA-N
storage temp.
0-5
Purity
90%
Country of Origin
US
Shipping Temperature
Ambient
Physical Form
Light Green Solid
Product Link
http://www.chemcia.com/product_fl02.asp?id=2608
MSDS
http://www.chemcia.com/product_fl02.asp?id=2608
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service