Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C12H316CE5E1

Chemcia Scientific, LLC

3-Hydroxy-benzamidine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
136.15
MDL number:

Iupac Name

3-hydroxybenzenecarboximidamide

Inchi Code

1S/C7H8N2O/c8-7(9)5-2-1-3-6(10)4-5/h1-4,10H,(H3,8,9)

InChI key

KXVXWFZZYKNVJS-UHFFFAOYSA-N

storage temp.

0-5C

Purity

96%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

Gray Solid

Product Link

http://www.chemcia.com/product_fl02.asp?id=1455

MSDS

http://www.chemcia.com/product_fl02.asp?id=1455


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service