C12H316CE465
Chemcia Scientific, LLC
3-Amino-5-bromo-thiophene-2-carboxylic acid methyl ester hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 3-amino-5-bromo-2-thiophenecarboxylate hydrochloride
Inchi Code
1S/C6H6BrNO2S.ClH/c1-10-6(9)5-3(8)2-4(7)11-5;/h2H,8H2,1H3;1H
InChI key
ORFVKWQXGICMRJ-UHFFFAOYSA-N
storage temp.
0-5C
Salt Data
HCl
Purity
97%
Country of Origin
US
Shipping Temperature
Ambient
Physical Form
Off-White Solid
Product Link
http://www.chemcia.com/product_fl02.asp?id=1214
MSDS
http://www.chemcia.com/product_fl02.asp?id=1214
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service